2-Fluor-5-methoxy-3-(trifluormethyl)benzylbromid 97%

Lieferant: Apollo Scientific

PC303036-10G PC303036-5G PC303036-1G
APOSPC303036-10GEA 0 EUR
APOSPC303036-10G APOSPC303036-5G APOSPC303036-1G
2-Fluor-5-methoxy-3-(trifluormethyl)benzylbromid 97%
2-Fluor-5-methoxy-3-(trifluormethyl)benzylbromid
Formel: C₉H₇BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD22201038
CAS-Nummer: 1373921-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR