2-Methoxy-5-(pentafluorothio)benzylamine 97%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-5-(pentafluorosulphur)benzylamine

PC303039-100MG
APOSPC303039-100MGEA 87 EUR
APOSPC303039-100MG
2-Methoxy-5-(pentafluorothio)benzylamine 97%
2-Methoxy-5-(pentafluorothio)benzylamine
Formel: C₈H₁₀F₅NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652492
CAS-Nummer: 1211516-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR