2-Methoxy-5-(trifluormethoxy)anilin 98%

Lieferant: Apollo Scientific
Gefahr

PC303041-10G PC303041-5G PC303041-1G PC303041-25G
APOSPC303041-10GEA 473 EUR
APOSPC303041-10G APOSPC303041-5G APOSPC303041-1G APOSPC303041-25G
2-Methoxy-5-(trifluormethoxy)anilin 98%
2-Methoxy-5-(trifluormethoxy)anilin
Formel: C₈H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832305
CAS-Nummer: 660848-57-7
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR