2-Methyl-5-(pentafluorothio)benzyl bromide 98%

Lieferant: Apollo Scientific

PC303046-1G PC303046-5G
APOSPC303046-1GEA 0 EUR
APOSPC303046-1G APOSPC303046-5G
2-Methyl-5-(pentafluorothio)benzyl bromide 98%
2-Methyl-5-(pentafluorothio)benzyl bromide
Formel: C₈H₈BrF₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD16652479
CAS-Nummer: 1240257-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR