2-Methyl-5-(trifluormethoxy)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC303049-10G PC303049-5G PC303049-1G
APOSPC303049-10GEA 560 EUR
APOSPC303049-10G APOSPC303049-5G APOSPC303049-1G
2-Methyl-5-(trifluormethoxy)benzylbromid
2-Methyl-5-(trifluormethoxy)benzylbromid
Formel: C₉H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18399774
CAS-Nummer: 1261573-60-7
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR