3-Chlor-5-(trifluormethyl)benzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC303061-5G PC303061-1G
APOSPC303061-5GEA 0 EUR
APOSPC303061-5G APOSPC303061-1G
3-Chlor-5-(trifluormethyl)benzylamin 98%
3-Chlor-5-(trifluormethyl)benzylamin
Formel: C₈H₇ClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD07786683
CAS-Nummer: 400771-41-7
UN: 2735
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR