3-Ethoxy-2,6-difluorphenol 98%

Lieferant: Apollo Scientific

PC303067-10G PC303067-5G PC303067-25G
APOSPC303067-10GEA 372 EUR
APOSPC303067-10G APOSPC303067-5G APOSPC303067-25G
3-Ethoxy-2,6-difluorphenol 98%
3-Ethoxy-2,6-difluorphenol
Formel: C₈H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11519373
CAS-Nummer: 1092461-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR