(3-Fluor-2-(trifluormethyl)phenyl)essigsäure 97%

Lieferant: Apollo Scientific

PC303068-1G
APOSPC303068-1GEA 203 EUR
APOSPC303068-1G
(3-Fluor-2-(trifluormethyl)phenyl)essigsäure 97%
(3-Fluor-2-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆F₄O₂
Molecular Weight: 222,14 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832251
CAS-Nummer: 897940-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR