3-Methyl-5-(pentafluorothio)benzyl bromide 98%
Lieferant: Apollo Scientific
Synonyme:
3-Methyl-5-(pentafluorosulphur)benzyl bromide
PC303071-5G
APOSPC303071-5GEA
0
EUR
APOSPC303071-5G
3-Methyl-5-(pentafluorothio)benzyl bromide 98%
3-Methyl-5-(pentafluorothio)benzyl bromide
Formel:
C₈H₈BrF₅S Molecular Weight: 311.11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652481 CAS-Nummer: 1240257-48-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...