5-Methyl-2-(trifluoromethoxy)benzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

PC303072-1G PC303072-250MG
APOSPC303072-1GEA 2210 EUR
APOSPC303072-1G APOSPC303072-250MG
5-Methyl-2-(trifluoromethoxy)benzenesulphonyl chloride 97%
5-Methyl-2-(trifluoromethoxy)benzenesulphonyl chloride
Formel: C₈H₆ClF₃O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD18399740
CAS-Nummer: 1261683-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR