4,5-Difluor-2-methoxybenzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC303074-5G
PC303074-1G
PC303074-25G
APOSPC303074-5GEA
203
EUR
APOSPC303074-5G
APOSPC303074-1G
APOSPC303074-25G
4,5-Difluor-2-methoxybenzylbromid 98%
4,5-Difluor-2-methoxybenzylbromid
Formel:
C₈H₇BrF₂O Schmelzpunkt: 40…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660220 CAS-Nummer: 886499-64-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...