4-Chlor-2-(trifluormethoxy)benzoesäure 98%

Lieferant: Apollo Scientific

PC303076-10G PC303076-5G PC303076-1G
APOSPC303076-10GEA 360 EUR
APOSPC303076-10G APOSPC303076-5G APOSPC303076-1G
4-Chlor-2-(trifluormethoxy)benzoesäure 98%
4-Chloro-2-(trifluoromethoxy)benzoic acid
Formel: C₈H₄ClF₃O₃
Molecular Weight: 240.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393801
CAS-Nummer: 1261605-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR