4-Chlor-2-(trifluormethoxy)benzoesäure 98%
Lieferant: Apollo Scientific
PC303076-10G
PC303076-5G
PC303076-1G
APOSPC303076-10GEA
360
EUR
APOSPC303076-10G
APOSPC303076-5G
APOSPC303076-1G
4-Chlor-2-(trifluormethoxy)benzoesäure 98%
4-Chloro-2-(trifluoromethoxy)benzoic acid
Formel:
C₈H₄ClF₃O₃ Molecular Weight: 240.56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393801 CAS-Nummer: 1261605-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...