4-Chloro-3-(trifluoromethoxy)benzamide 98%

Lieferant: Apollo Scientific

PC303079-1G
APOSPC303079-1GEA 69.6 EUR
APOSPC303079-1G
4-Chloro-3-(trifluoromethoxy)benzamide 98%
4-Chloro-3-(trifluoromethoxy)benzamide
Formel: C₈H₅ClF₃NO₂
Molecular Weight: 239,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660246
CAS-Nummer: 886500-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR