4-Fluoro-3-methyl-5-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific
Achtung

PC303090-10G PC303090-5G PC303090-1G
APOSPC303090-10GEA 613 EUR
APOSPC303090-10G APOSPC303090-5G APOSPC303090-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)aniline 98%
4-Fluoro-3-methyl-5-(trifluoromethyl)aniline
Formel: C₈H₇F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD22201067
CAS-Nummer: 1373920-72-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR