5-Chlor-2-(trifluormethyl)benzylamin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC303095-1G
APOSPC303095-1GEA
0
EUR
APOSPC303095-1G
5-Chlor-2-(trifluormethyl)benzylamin 95%
5-Chlor-2-(trifluormethyl)benzylamin
Formel:
C₈H₇ClF₃N Molecular Weight: 209,60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832359 CAS-Nummer: 942319-05-3 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...