2-Fluoro-4,6-bis(trifluoromethyl)phenylacetonitrile 97%
Lieferant: Apollo Scientific
PC303102-1G
APOSPC303102-1GEA
215
EUR
APOSPC303102-1G
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetonitrile 97%
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetonitrile
Formel:
C₁₀H₄F₇N Schmelzpunkt: 34…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258674 CAS-Nummer: 1017778-50-5 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...