5-Methyl-2-(trifluoromethyl)benzamide

Lieferant: Apollo Scientific

PC303103-10G PC303103-5G
APOSPC303103-10GEA 0 EUR
APOSPC303103-10G APOSPC303103-5G
5-Methyl-2-(trifluoromethyl)benzamide
5-Methyl-2-(trifluoromethyl)benzamide
Formel: C₉H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06660293
CAS-Nummer: 886502-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR