2-Chlor-6-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific

PC303105-1G
APOSPC303105-1GEA 0 EUR
APOSPC303105-1G
2-Chlor-6-(trifluormethyl)benzonitril 98%
2-Chlor-6-(trifluormethyl)benzonitril
Formel: C₈H₃ClF₃N
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412191
CAS-Nummer: 129604-28-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR