4-Ethoxy-2-fluorobenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC303114-1G
APOSPC303114-1GEA 94.4 EUR
APOSPC303114-1G
4-Ethoxy-2-fluorobenzoic acid 98%
4-Ethoxy-2-fluorobenzoic acid
Formel: C₉H₉FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13196593
CAS-Nummer: 1206593-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR