4-Methoxy-2-(trifluormethyl)benzylamin 98%

Lieferant: Apollo Scientific

PC303115-1G
APOSPC303115-1GEA 0 EUR
APOSPC303115-1G
4-Methoxy-2-(trifluormethyl)benzylamin 98%
4-Methoxy-2-(trifluormethyl)benzylamin
Formel: C₉H₁₀F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09025405
CAS-Nummer: 916420-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR