2-Chlor-4-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC303117-1G PC303117-5G PC303117-10G
APOSPC303117-1GEA 203 EUR
APOSPC303117-1G APOSPC303117-5G APOSPC303117-10G
2-Chlor-4-(trifluormethyl)benzylbromid 98%
2-Chlor-4-(trifluormethyl)benzylbromid
Formel: C₈H₅BrClF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14155638
CAS-Nummer: 279252-26-5
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR