2-Fluoro-4,6-bis(trifluoromethyl)phenylacetic acid 98%

Lieferant: Apollo Scientific
Achtung

PC303118-1G
APOSPC303118-1GEA 0 EUR
APOSPC303118-1G
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetic acid
Formel: C₁₀H₅F₇O₂
Schmelzpunkt: 99…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258675
CAS-Nummer: 1017778-54-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR