2-Methoxy-5-(pentafluorosulphur)phenylacetic acid

Lieferant: Apollo Scientific

PC303125-1G
APOSPC303125-1GEA 339 EUR
APOSPC303125-1G
2-Methoxy-5-(pentafluorosulphur)phenylacetic acid
2-Methoxy-5-(pentafluorosulphur)phenylacetic acid
Formel: C₉H₉F₅O₃S
Molecular Weight: 292,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652490
CAS-Nummer: 1240257-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR