4-n-Butoxy-2,6-difluoroaniline 97%

Lieferant: Apollo Scientific

Synonyme: 4-Butoxy-2,6-difluoroaniline

PC303126-1G
APOSPC303126-1GEA 271 EUR
APOSPC303126-1G
4-n-Butoxy-2,6-difluoroaniline 97%
4-n-Butoxy-2,6-difluoroaniline
Formel: C₁₀H₁₃F₂NO
Molecular Weight: 201,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201028
CAS-Nummer: 1373920-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR