2,5-Dichloro-3-(trifluoromethyl)aniline 97%

Lieferant: Apollo Scientific
Gefahr

PC303132-10G PC303132-5G PC303132-1G
APOSPC303132-10GEA 485 EUR
APOSPC303132-10G APOSPC303132-5G APOSPC303132-1G
2,5-Dichloro-3-(trifluoromethyl)aniline 97%
2,5-Dichloro-3-(trifluoromethyl)aniline
Formel: C₇H₄Cl₂F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD27986858
CAS-Nummer: 103995-81-9
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR