2-Methoxy-4-(trifluormethoxy)anilin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC303133-1G
APOSPC303133-1GEA
0
EUR
APOSPC303133-1G
2-Methoxy-4-(trifluormethoxy)anilin 98%
2-Methoxy-4-(trifluormethoxy)anilin
Formel:
C₈H₈F₃NO₂ Molecular Weight: 207.15 g/mol Schmelzpunkt: 37…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832304 CAS-Nummer: 131395-23-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...