2-Methyl-4-trifluoromethoxy-DL-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-4-(trifluoromethoxy)-DL-phenylalanine
PC303135-1G
APOSPC303135-1GEA
159
EUR
APOSPC303135-1G
2-Methyl-4-trifluoromethoxy-DL-phenylalanine
2-Methyl-4-trifluoromethoxy-DL-phenylalanine
Formel:
C₁₁H₁₂F₃NO₃ Molecular Weight: 263.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201062 CAS-Nummer: 1706428-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...