4-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303137-1G PC303137-5G
APOSPC303137-1GEA 0 EUR
APOSPC303137-1G APOSPC303137-5G
4-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%
4-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27986860
CAS-Nummer: 1706436-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR