2,4-Difluor-3-methoxybenzylamin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC303140-1G
APOSPC303140-1GEA
0
EUR
APOSPC303140-1G
2,4-Difluor-3-methoxybenzylamin 97%
2,4-Difluor-3-methoxybenzylamin
Formel:
C₈H₉F₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115935 CAS-Nummer: 717094-51-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...