2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC303145-1G
PC303145-5G
APOSPC303145-1GEA
339
EUR
APOSPC303145-1G
APOSPC303145-5G
2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde 98%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde
Formel:
C₉H₆F₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054185 CAS-Nummer: 1706458-33-4 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...