2-Fluoro-5-methyl-3-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303146-1G
APOSPC303146-1GEA 339 EUR
APOSPC303146-1G
2-Fluoro-5-methyl-3-(trifluoromethyl)benzoic acid 97%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
Molecular Weight: 222,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054184
CAS-Nummer: 1706435-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR