2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC303154-1G
PC303154-250MG
APOSPC303154-1GEA
507
EUR
APOSPC303154-1G
APOSPC303154-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde
Formel:
C₉H₆F₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD28103358 CAS-Nummer: 1824270-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...