2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC303154-1G PC303154-250MG
APOSPC303154-1GEA 507 EUR
APOSPC303154-1G APOSPC303154-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde
Formel: C₉H₆F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD28103358
CAS-Nummer: 1824270-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR