5-Chloro-2-methyl-4-(trifluoromethoxy)aniline 98%

Lieferant: Apollo Scientific

PC303162-5G PC303162-1G
APOSPC303162-5GEA 574 EUR
APOSPC303162-5G APOSPC303162-1G
5-Chloro-2-methyl-4-(trifluoromethoxy)aniline 98%
5-Chloro-2-methyl-4-(trifluoromethoxy)aniline
Formel: C₈H₇ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD27986859
CAS-Nummer: 1706461-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR