2,6-Dichloro-3-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303178-1G
APOSPC303178-1GEA 676 EUR
APOSPC303178-1G
2,6-Dichloro-3-(trifluoromethyl)benzenesulphonyl chloride
2,6-Dichloro-3-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂Cl₃F₃O₂S
Molecular Weight: 313,509 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054229
CAS-Nummer: 1706458-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR