2-Chloro-3,6-difluorobenzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303180-1G
APOSPC303180-1GEA
676
EUR
APOSPC303180-1G
2-Chloro-3,6-difluorobenzenesulphonyl chloride
2-Chloro-3,6-difluorobenzenesulphonyl chloride
Formel:
C₆H₂Cl₂F₂O₂S Molecular Weight: 247,047 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922610 CAS-Nummer: 1208074-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...