3-Chloro-2-fluoro-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303186-1G
APOSPC303186-1GEA 676 EUR
APOSPC303186-1G
3-Chloro-2-fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
3-Chloro-2-fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂Cl₂F₄O₂S
Molecular Weight: 297,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054228
CAS-Nummer: 1706458-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR