3-Chloro-2-fluoro-6-(trifluoromethyl)benzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303187-1G
APOSPC303187-1GEA
1120
EUR
APOSPC303187-1G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzenesulphonyl chloride
3-Chloro-2-fluoro-6-(trifluoromethyl)benzenesulphonyl chloride
Formel:
C₇H₂Cl₂F₄O₂S Molecular Weight: 297,054 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054231 CAS-Nummer: 1706461-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...