4-Ethoxy-3-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303189-1G
APOSPC303189-1GEA 676 EUR
APOSPC303189-1G
4-Ethoxy-3-(trifluoromethyl)benzenesulphonyl chloride
4-Ethoxy-3-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₉H₈ClF₃O₃S
Molecular Weight: 288,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054232
CAS-Nummer: 1706446-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR