2,3-Dichloro-6-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303190-1G
APOSPC303190-1GEA 731 EUR
APOSPC303190-1G
2,3-Dichloro-6-(trifluoromethyl)benzenesulphonyl chloride
2,3-Dichloro-6-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂Cl₃F₃O₂S
Molecular Weight: 313,509 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054221
CAS-Nummer: 1706446-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR