2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific

PC303193-1G PC303193-250MG
APOSPC303193-1GEA 0 EUR
APOSPC303193-1G APOSPC303193-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl bromide
Formel: C₉H₇BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD28103367
CAS-Nummer: 1824048-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR