2,3-Difluoro-4-methoxybenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303198-1G
APOSPC303198-1GEA 788 EUR
APOSPC303198-1G
2,3-Difluoro-4-methoxybenzenesulphonyl chloride
2,3-Difluoro-4-methoxybenzenesulphonyl chloride
Formel: C₇H₅ClF₂O₃S
Molecular Weight: 242,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17282234
CAS-Nummer: 1341660-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR