2,3-Difluoro-4-methylbenzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303199-1G
APOSPC303199-1GEA
788
EUR
APOSPC303199-1G
2,3-Difluoro-4-methylbenzenesulphonyl chloride
2,3-Difluoro-4-methylbenzenesulphonyl chloride
Formel:
C₇H₅ClF₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054217 CAS-Nummer: 1698682-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...