2-Methoxy-5-(trifluoromethoxy)benzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

PC303204-1G PC303204-250MG
APOSPC303204-1GEA 0 EUR
APOSPC303204-1G APOSPC303204-250MG
2-Methoxy-5-(trifluoromethoxy)benzenesulphonyl chloride 97%
2-Methoxy-5-(trifluoromethoxy)benzenesulphonyl chloride
Formel: C₈H₆ClF₃O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD13176696
CAS-Nummer: 1010422-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR