4-Chloro-3,5-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC303208-1G
APOSPC303208-1GEA 788 EUR
APOSPC303208-1G
4-Chloro-3,5-difluorobenzenesulphonyl chloride
4-Chloro-3,5-difluorobenzenesulphonyl chloride
Formel: C₆H₂Cl₂F₂O₂S
Molecular Weight: 247,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054249
CAS-Nummer: 1706435-19-9
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR