2-Chloro-6-fluoro-3-methylbenzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303213-1G
APOSPC303213-1GEA
845
EUR
APOSPC303213-1G
2-Chloro-6-fluoro-3-methylbenzenesulphonyl chloride
2-Chloro-6-fluoro-3-methylbenzenesulphonyl chloride
Formel:
C₇H₅Cl₂FO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054213 CAS-Nummer: 1706430-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...