2-Fluoro-4-methyl-3-(trifluoromethyl)benzylamine

Lieferant: Apollo Scientific

PC303216-1G
APOSPC303216-1GEA 845 EUR
APOSPC303216-1G
2-Fluoro-4-methyl-3-(trifluoromethyl)benzylamine
2-Fluoro-4-methyl-3-(trifluoromethyl)benzylamine
Formel: C₉H₉F₄N
Molecular Weight: 207,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28103370
CAS-Nummer: 1824271-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR