6-Chloro-2-fluoro-3-methylbenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303219-1G
APOSPC303219-1GEA 845 EUR
APOSPC303219-1G
6-Chloro-2-fluoro-3-methylbenzenesulphonyl chloride
6-Chloro-2-fluoro-3-methylbenzenesulphonyl chloride
Formel: C₇H₅Cl₂FO₂S
Molecular Weight: 243,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054215
CAS-Nummer: 1706461-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR