2-Methoxy-5-(pentafluorosulphur)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303221-1G
APOSPC303221-1GEA 866 EUR
APOSPC303221-1G
2-Methoxy-5-(pentafluorosulphur)benzenesulphonyl chloride
2-Methoxy-5-(pentafluorosulphur)benzenesulphonyl chloride
Formel: C₇H₆ClF₅O₃S₂
Molecular Weight: 332,696 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054237
CAS-Nummer: 1706447-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR